Order Entry
Czech Republic
ContactUsLinkComponent
7-Aminothieno[2,3-b]pyrazine 95%
Toxic
7-Aminothieno[2,3-b]pyrazine 95%
Katalogové číslo APOSOR9221-250MG
Supplier:  Apollo Scientific
CAS Number:  
Toxic
7-Aminothieno[2,3-b]pyrazine 95%
Katalogové číslo APOSOR9221-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :151.189 g/mol
Melting Pt:87.5…89.5 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:59944-75-1
Formula:C₆H₅N₃S
MDL Number:MFCD08271970