
(1R,4R)-2-Methyl-2,5-diazabicyclo[2.2.1]heptane dihydrobromide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:125224-64-8
:
Formula:C₆H₁₂N₂·2BrH
MDL Number:MFCD13195528
:273.997 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together