Order Entry
Czech Republic
ContactUsLinkComponent
(R)-3-Methoxypiperidine hydrochloride 98%
(R)-3-Methoxypiperidine hydrochloride 98%
Catalog # APOSOR921704-1G
Supplier:  Apollo Scientific
CAS Number:  
(R)-3-Methoxypiperidine hydrochloride 98%
Catalog # APOSOR921704-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :151.634 g/mol
Storage Temperature:Ambient
CAS Number:688809-95-2
Formula:C₆H₁₃NO·ClH
MDL Number:MFCD17015899