3-(Aminomethyl)-4,6-dimethyl-1,2-dihydropyridin-2-one 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:771579-27-2
:
Formula:C₈H₁₂N₂O
MDL Number:MFCD06213260
:152.194 g/mol
Storage Temperature:Refrigerator
Frequently Bought Together