Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 3-methyl-1H-pyrrole-2-carboxylate 95%
Methyl 3-methyl-1H-pyrrole-2-carboxylate 95%
Catalog # APOSOR921497-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 3-methyl-1H-pyrrole-2-carboxylate 95%
Catalog # APOSOR921497-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :139.152 g/mol
Storage Temperature:Ambient
CAS Number:40611-69-6
Formula:C₇H₉NO₂
MDL Number:MFCD09754196