Order Entry
Czech Republic
ContactUsLinkComponent
 
N-[2-(tert-Butyldimethylsilyloxy)ethyl]methylamine 97%
Catalog # APOSOR920911-1G
Supplier:  Apollo Scientific
CAS Number:  
N-[2-(tert-Butyldimethylsilyloxy)ethyl]methylamine 97%
Catalog # APOSOR920911-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :189.371 g/mol
Storage Temperature:Ambient
CAS Number:204580-28-9
Formula:C₉H₂₃NOSi
MDL Number:MFCD20662141