
7-Methyl-2,7-diazaspiro[3.5]nonane dihydrochloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1588441-26-2
:
Formula:C₈H₁₆N₂·2ClH
MDL Number:MFCD26743659
:213.148 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together