4-Oxa-7-azaspiro[2.5]octane hydrochloride
Catalog # APOSOR920787-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:149.619 g/mol
Storage Temperature:Ambient
CAS Number:1427195-23-0
Formula:C₆H₁₁NO·ClH
MDL Number:MFCD21606609