
1-Oxa-8-azaspiro[4.5]decan-3-one hydrochloride 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:133382-42-0
:
Formula:C₈H₁₃NO₂·ClH
MDL Number:MFCD13193037
:191.655 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together