1,4-Dioxa-spiro[4.5]dec-8-ylamine hydrochloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:155137-22-7
:
Formula:C₈H₁₅NO₂·ClH
MDL Number:MFCD26793861
:193.671 g/mol
Storage Temperature:Ambient
Frequently Bought Together