Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
7-Oxa-4-azaspiro[2.5]octane hydrochloride
7-Oxa-4-azaspiro[2.5]octane hydrochloride 98%
7-Oxa-4-azaspiro[2.5]octane hydrochloride 98%
7-Oxa-4-azaspiro[2.5]octane hydrochloride 98%
APOSOR918050-100MG
:
Apollo Scientific
:
7-Oxa-4-azaspiro[2.5]octane hydrochloride 98%
APOSOR918050-100MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 mg
Seq:
2440
:
:
149.619 g/mol
:
Freezer
:
218595-22-3
:
C₆H₁₁NO·ClH
:
MFCD18914346