Order Entry
Czech Republic
ContactUsLinkComponent
5-Methyl-5-propyl-1,3-dioxan-2-one 95%
5-Methyl-5-propyl-1,3-dioxan-2-one 95%
Catalog # APOSOR917585-5G
Supplier:  Apollo Scientific
CAS Number:  
5-Methyl-5-propyl-1,3-dioxan-2-one 95%
Catalog # APOSOR917585-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :158.195 g/mol
Storage Temperature:Ambient
CAS Number:7148-50-7
Formula:C₈H₁₄O₃
MDL Number:MFCD00023865