Order Entry
Czech Republic
ContactUsLinkComponent
L-4-Methoxymandelic acid 95%
L-4-Methoxymandelic acid 95%
Catalog # APOSOR916843-1G
Supplier:  Apollo Scientific
CAS Number:  
L-4-Methoxymandelic acid 95%
Catalog # APOSOR916843-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :182.173 g/mol
Storage Temperature:Ambient
CAS Number:75172-66-6
Formula:C₉H₁₀O₄
MDL Number:MFCD00754498