
3-(1,3-Dioxolan-2-yl)thiophene-2-sulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:103011-38-7
:
Formula:C₇H₇ClO₄S₂
MDL Number:MFCD08271969
:254.711 g/mol
Melting Pt:93…94 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together