(S)-3,3,6,6-Tetramethyltetrahydropyrrolo[1,2-c]oxazol-5(3H)-one 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:156088-45-8
:
Formula:C₁₀H₁₇NO₂
MDL Number:MFCD19382367
:183.247 g/mol
Storage Temperature:Ambient
Frequently Bought Together