(2S)-1-(1-Methylethyl)azetidine-2-carboxylic acid
Catalog # APOSOR913830-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:143.184 g/mol
Storage Temperature:Ambient
CAS Number:255882-98-5
Formula:C₇H₁₃NO₂
MDL Number:MFCD11053659