Order Entry
Czech Republic
ContactUsLinkComponent
2-Chloro-4-hydroxy-benzoic acid methyl ester 98%
2-Chloro-4-hydroxy-benzoic acid methyl ester 98%
Catalog # APOSOR913032-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-4-hydroxy-benzoic acid methyl ester 98%
Catalog # APOSOR913032-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :186.592 g/mol
Storage Temperature:Ambient
CAS Number:104253-44-3
Formula:C₈H₇ClO₃
MDL Number:MFCD04117954