Order Entry
Czech Republic
ContactUsLinkComponent
4-(N-methylacetamido)benzoic acid 97%
4-(N-methylacetamido)benzoic acid 97%
Catalog # APOSOR911241-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(N-methylacetamido)benzoic acid 97%
Catalog # APOSOR911241-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :193.199 g/mol
Storage Temperature:Ambient
CAS Number:26961-99-9
Formula:C₁₀H₁₁NO₃
MDL Number:MFCD20542852