Order Entry
Czech Republic
ContactUsLinkComponent
(2E)-3,4,4-trimethylpent-2-enamide 97%
(2E)-3,4,4-trimethylpent-2-enamide 97%
Catalog # APOSOR908070-1G
Supplier:  Apollo Scientific
CAS Number:  
(2E)-3,4,4-trimethylpent-2-enamide 97%
Catalog # APOSOR908070-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :141.211 g/mol
Storage Temperature:Ambient
CAS Number:860546-40-3
Formula:C₈H₁₅NO
MDL Number:MFCD19206375