Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
trans-4-(4-Morpholinyl)cyclohexanol 98%

trans-4-(4-Morpholinyl)cyclohexanol 98%

New Product
Supplier:   Apollo Scientific
trans-4-(4-Morpholinyl)cyclohexanol 98%
Supplier:  Apollo Scientific
New Product
Certificates
trans-4-(4-Morpholinyl)cyclohexanol 98%

3 Options Available Below

Product Details & Documents
Synonyms:  trans-4-Morpholinocyclohexanol
  • CAS Number:1228947-14-5
  • Formula:C₁₀H₁₉NO₂
  • MDL Number:MFCD20259655
  • Molecular Weight:185.263 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 1 560,00 Kč
    1 560,00 Kč
    1 * 250 mg
       1 560,00 Kč

    Specifications:
    New Product
    New Product
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 3 080,00 Kč
    3 080,00 Kč
    1 * 1 g
       3 080,00 Kč

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 9 190,00 Kč
    9 190,00 Kč
    1 * 5 g
       9 190,00 Kč

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.