Order Entry
Czech Republic
ContactUsLinkComponent
2-(3-Methoxyphenyl)pyrrolidine 95%
2-(3-Methoxyphenyl)pyrrolidine 95%
Catalog # APOSOR905430-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(3-Methoxyphenyl)pyrrolidine 95%
Catalog # APOSOR905430-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :177.243 g/mol
Storage Temperature:Ambient
CAS Number:103861-77-4
Formula:C₁₁H₁₅NO
MDL Number:MFCD02663445