
N,N-Diethyl-2-(1-naphthyloxy)propanamide
Catalog # APOSOR904188-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:271.354 g/mol
Melting Pt:73…78 °C
:GHS07
Storage Temperature:Ambient
CAS Number:15299-99-7
Formula:C₁₇H₂₁NO₂
MDL Number:MFCD00055354