Order Entry
Czech Republic
ContactUsLinkComponent
2-Methyl-4-(4-methylphenyl)-1,3-thiazole 95%
Irritant
2-Methyl-4-(4-methylphenyl)-1,3-thiazole 95%
Catalog # APOSOR903268-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Methyl-4-(4-methylphenyl)-1,3-thiazole 95%
Catalog # APOSOR903268-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :189.277 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:66047-73-2
Formula:C₁₁H₁₁NS
MDL Number:MFCD00662551