Order Entry
Czech Republic
ContactUsLinkComponent
2-(4-Methoxyphenyl)-2-methylpropylamine 98%
2-(4-Methoxyphenyl)-2-methylpropylamine 98%
Catalog # APOSOR903208-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Methoxyphenyl)-2-methylpropylamine 98%
Catalog # APOSOR903208-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :179.259 g/mol
Storage Temperature:Ambient
CAS Number:51558-25-9
Formula:C₁₁H₁₇NO
MDL Number:MFCD01937048