Order Entry
Czech Republic
ContactUsLinkComponent
2,2-Dimethoxyethyl isocyanide 95%
2,2-Dimethoxyethyl isocyanide 95%
Catalog # APOSOR903045-1G
Supplier:  Apollo Scientific
CAS Number:  
2,2-Dimethoxyethyl isocyanide 95%
Catalog # APOSOR903045-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :115.13 g/mol
Storage Temperature:Ambient
CAS Number:277300-82-0
Formula:C₅H₉NO₂
MDL Number:MFCD02664543