
tert-Butyl (2-chloropyridin-4-yl)methylcarbamate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:916210-27-0
:
Formula:C₁₁H₁₅ClN₂O₂
MDL Number:MFCD09028165
:242.702 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together