Order Entry
Czech Republic
ContactUsLinkComponent
2-(3-Bromopyridin-2-yl)acetonitrile 98%
2-(3-Bromopyridin-2-yl)acetonitrile 98%
Catalog # APOSOR901848-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(3-Bromopyridin-2-yl)acetonitrile 98%
Catalog # APOSOR901848-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :197.032 g/mol
Storage Temperature:Ambient
CAS Number:122851-60-9
Formula:C₇H₅BrN₂
MDL Number:MFCD09999977