Order Entry
Czech Republic
ContactUsLinkComponent
2-[(4-Chlorophenyl)(hydroxy)methyl]prop-2-enenitrile 95%
2-[(4-Chlorophenyl)(hydroxy)methyl]prop-2-enenitrile 95%
Catalog # APOSOR900787-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[(4-Chlorophenyl)(hydroxy)methyl]prop-2-enenitrile 95%
Catalog # APOSOR900787-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :193.63 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₈ClNO
MDL Number:MFCD11130359