(3Z)-4-(Cyclopropylamino)pent-3-en-2-one 98%
Catalog # APOSOR900331-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:139.195 g/mol
Storage Temperature:Ambient
CAS Number:1314996-30-9
Formula:C₈H₁₃NO
MDL Number:MFCD19159684