
2-Methyl-4-phenyl-1,3-thiazole-5-carboxaldehyde 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:857284-11-8
:
Formula:C₁₁H₉NOS
MDL Number:MFCD08690246
:203.26 g/mol
Melting Pt:72…75 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together