
Methyl 5-(chlorosulphonyl)-2-methyl-3-furoate 95+%
Catalog # APOSOR8121-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:238.645 g/mol
Melting Pt:35…38 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05
CAS Number:306936-35-6
Formula:C₇H₇ClO₅S
MDL Number:MFCD02180742