
2,5-Diethoxy-4-(morpholin-4-yl)aniline
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:51963-82-7
:
Formula:C₁₄H₂₂N₂O₃
MDL Number:MFCD02052680
:266.336 g/mol
Melting Pt:93…96 °C
Density:1.13 g/cm³ (25 °C)
EINECS:257-562-7
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together