
5-Chloro-1-methyl-1H-pyrazole-4-carbonyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:110763-09-2
:
Formula:C₅H₄Cl₂N₂O
MDL Number:MFCD02179947
:179.004 g/mol
Melting Pt:50…52 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together