Order Entry
Czech Republic
ContactUsLinkComponent
3-(1-Methyl-1H-pyrazol-3-yl)aniline
Irritant
3-(1-Methyl-1H-pyrazol-3-yl)aniline
Catalog # APOSOR8034-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-(1-Methyl-1H-pyrazol-3-yl)aniline
Catalog # APOSOR8034-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Melting Pt:116…119 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
 :173.214 g/mol
 :GHS07
CAS Number:175202-37-6
Formula:C₁₀H₁₁N₃
MDL Number:MFCD00084885