Order Entry
Czech Republic
ContactUsLinkComponent
2-(2-Amino-4-methylthien-3-yl)-1,3-benzothiazole 97%
Irritant
2-(2-Amino-4-methylthien-3-yl)-1,3-benzothiazole 97%
Catalog # APOSOR8011-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(2-Amino-4-methylthien-3-yl)-1,3-benzothiazole 97%
Catalog # APOSOR8011-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :246.351 g/mol
Melting Pt:110…112 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:232941-00-3
Formula:C₁₂H₁₀N₂S₂
MDL Number:MFCD02179980