

2-(Thien-2-yl)aniline hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1087723-62-3
:
Formula:C₁₀H₉NS·ClH
MDL Number:MFCD07365346
:211.711 g/mol
Melting Pt:210…212 °C (lit.)
UN:2811
ADR:6.1,III
:GHS05
:GHS06
Storage Temperature:Ambient
Frequently Bought Together