
2-(5-Methylfur-2-yl)aniline hydrochloride
Catalog # APOSOR7535-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:209.672 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:329187-42-0
Formula:C₁₁H₁₁NO·ClH
MDL Number:MFCD01191726