Order Entry
Czech Republic
ContactUsLinkComponent
2'-Methyl[1,1'-biphenyl]-2-amine hydrochloride
Irritant
2'-Methyl[1,1'-biphenyl]-2-amine hydrochloride
Catalog # APOSOR7490-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2'-Methyl[1,1'-biphenyl]-2-amine hydrochloride
Catalog # APOSOR7490-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :219.71 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:1203-41-4
Formula:C₁₃H₁₃N·ClH
MDL Number:MFCD06739417