Order Entry
Czech Republic
ContactUsLinkComponent
Methyl 3'-amino-[1,1'-biphenyl]-3-carboxylate hydrochloride 98%
Irritant
Methyl 3'-amino-[1,1'-biphenyl]-3-carboxylate hydrochloride 98%
Catalog # APOSOR7484-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl 3'-amino-[1,1'-biphenyl]-3-carboxylate hydrochloride 98%
Catalog # APOSOR7484-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :263.719 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:343985-94-4
Formula:C₁₄H₁₃NO₂·ClH
MDL Number:MFCD07366482