

5-(3-Methoxyphenyl)-2-furoic acid
Catalog # APOSOR7479-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:218.205 g/mol
Melting Pt:169…170 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
:GHS07
CAS Number:54022-96-7
Formula:C₁₂H₁₀O₄
MDL Number:MFCD05022489