3-(4-Bromophenyl)cyclobutan-1-one
Catalog # APOSOR7045-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:225.082 g/mol
Storage Temperature:Ambient
CAS Number:254892-91-6
Formula:C₁₀H₉BrO
MDL Number:MFCD04038979