
2-Bromo-3-chlorobenzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:56961-26-3
:
Formula:C₇H₄BrClO₂
MDL Number:MFCD00672931
:235.462 g/mol
Melting Pt:143…146 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together