
1,5-Dioxa-9-azaspiro[5.5]undecane, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:180-94-9
:
Formula:C₈H₁₅NO₂
MDL Number:MFCD00040749
:157.21 g/mol
Boiling Pt:65…67 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together