Order Entry
Czech Republic
ContactUsLinkComponent
2-(Thien-2-yl)benzylamine hydrochloride 95%
Irritant
2-(Thien-2-yl)benzylamine hydrochloride 95%
Catalog # APOSOR6806-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(Thien-2-yl)benzylamine hydrochloride 95%
Catalog # APOSOR6806-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :225.738 g/mol
Melting Pt:186.5…188.5 °C
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
 :GHS07
CAS Number:863991-95-1
Formula:C₁₁H₁₁NS·ClH
MDL Number:MFCD07772821