Order Entry
Czech Republic
ContactUsLinkComponent
 
Methyl 4-amino-3-bromo-5-chlorobenzoate 97%
Catalog # APOSOR6764-250MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl 4-amino-3-bromo-5-chlorobenzoate 97%
Catalog # APOSOR6764-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :264.504 g/mol
Melting Pt:125…127 °C
Storage Temperature:Ambient
CAS Number:914636-88-7
Formula:C₈H₇BrClNO₂
MDL Number:MFCD08436084