Order Entry
Czech Republic
ContactUsLinkComponent
4-Bromo-2-methoxybenzenesulphonyl chloride 97%
Corrosive
4-Bromo-2-methoxybenzenesulphonyl chloride 97%
Catalog # APOSOR61543-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
4-Bromo-2-methoxybenzenesulphonyl chloride 97%
Catalog # APOSOR61543-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :285.543 g/mol
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
CAS Number:145915-29-3
Formula:C₇H₆BrClO₃S
MDL Number:MFCD11847575