
2-Ethoxy-5-methoxybenzeneboronic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:957065-85-9
:
Formula:C₉H₁₃BO₄
MDL Number:MFCD09878346
:196.008 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together