
2-Acetamido-1-amino-1,2-dideoxy-β-D-glucopyranose 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4229-38-3
:
Formula:C₈H₁₆N₂O₅
MDL Number:MFCD00057751
:220.223 g/mol
Melting Pt:103…112 °C (dec.)
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together