
8-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,9b-tetrahydrobenzo[c]thieno[2,1-e]isothiazole 4-oxide 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2169311-10-6
:
Formula:C₁₆H₂₂BNO₃S
MDL Number:MFCD31629920
:319.227 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together