
Methyl 1,2,3,9b-tetrahydro-4λ4-benzo[c]thieno[2,1-e]isothiazole-8-carboxylate 4-oxide 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2169310-99-8
:
Formula:C₁₂H₁₃NO₃S
MDL Number:MFCD31629916
:251.302 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together